Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ S100A8 Recombinant Rabbit Monoclonal Antibody (203)
GREENER_CHOICE

Catalog No. PIMA536007
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA536007 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA536007 Supplier Invitrogen™ Supplier No. MA536007
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

S100A8 Recombinant Monoclonal Antibody for Western Blot, IP

The protein encoded by this gene is a member of the S100 family of proteins containing 2 EF-hand calcium-binding motifs. S100 proteins are localized in the cytoplasm and/or nucleus of a wide range of cells, and involved in the regulation of a number of cellular processes such as cell cycle progression and differentiation. S100 genes include at least 13 members which are located as a cluster on chromosome 1q21. This protein may function in the inhibition of casein kinase and as a cytokine. Altered expression of this protein is associated with the disease cystic fibrosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen S100A8
Applications Immunoprecipitation, Western Blot
Classification Recombinant Monoclonal
Clone 203
Conjugate Unconjugated
Formulation PBS with no preservative
Gene S100A8
Gene Accession No. P05109
Gene Alias 60B8AG; AI323541; B8Ag; CAGA; CAGB; calgranulin A; calgranulin B; calgranulin-A; Calgranulin-B; calprotectin L1H subunit; Calprotectin L1L subunit; CFAg; CGLA; CGLB; chemotactic cytokine CP-10; chemotactic S100 protein; CP-10; Cystic fibrosis antigen; L1Ag; leukocyte L1 complex heavy chain; Leukocyte L1 complex light chain; LIAG; MA387; MAC387; MIF; Migration inhibitory factor-related protein 14; migration inhibitory factor-related protein 8; MRP14; MRP-14; MRP8; MRP-8; NIF; P14; p8; Pro-inflammatory S100 cytokine; protein S100-A8; Protein S100-A9; S100 A8; S100 calcium binding protein A8; S100 calcium binding protein A8 (calgranulin A); S100 calcium binding protein A9; S100 calcium-binding protein A8; S100 calcium-binding protein A8 (calgranulin A); S100 calcium-binding protein A9; S100 calcium-binding protein A9 (calgranulin B); S100A8; S100A9; Urinary stone protein band A
Gene Symbols S100A8
Host Species Rabbit
Immunogen Recombinant Human S100A8 Protein.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6279
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.