Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
S100A8/S100A9 Heterodimer Antibody (900027), DyLight 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB45702K
Description
S100A8/S100A9 Heterodimer Monoclonal antibody specifically detects S100A8/S100A9 Heterodimer in Human samples. It is validated for ELISA, Immunohistochemistry, Flow Cytometry (Intracellular Staining), CyTOFSpecifications
S100A8/S100A9 Heterodimer | |
Monoclonal | |
DyLight 488 | |
50mM Sodium Borate | |
E. coli-derived recombinant human S100A8/S100A9 heterodimer, Met1-Glu93 (S100A8) & Thr2-Pro114 (S100A9), Accession # P05109 (S100A8) and P06702 (S100A9) | |
0.1 mL | |
Primary | |
Store at 4°C in the dark. | |
IgG1 |
ELISA, Immunohistochemistry, Flow Cytometry (Intracellular Staining), CyTOF | |
900027 | |
ELISA, Immunohistochemistry, Intracellular Staining by Flow Cytometry, CyTOF-ready | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction