Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
S100A9 Antibody (018), HRP

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289817H
Description
S100A9 Monoclonal specifically detects S100A9 in Human samples. It is validated for Western Blot, ELISA.Specifications
S100A9 | |
Monoclonal | |
HRP | |
CAGBMigration inhibitory factor-related protein 14, calgranulin B, calgranulin-B, Calprotectin L1H subunit, CFAGMRP-14,60B8AG, CGLB, L1AG, LIAG, MAC387, MIF, MRP14Leukocyte L1 complex heavy chain, NIF, P14, protein S100-A9, S100 calcium binding protein A9, S100 calcium binding protein A9 (calgranulin B), S100 calcium-binding protein A9, S100 calcium-binding protein A9 (calgranulin B) | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human S100A9 (NP_002956.1; Met 1-Pro 114) . | |
0.1 mL | |
Cancer, Immunology, Innate Immunity, Signal Transduction, Tyrosine Kinases | |
6280 | |
Human | |
Purified |
Western Blot, ELISA | |
18 | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
No cross-reactivity in ELISA with: Human S100A13 Human S100A1 Human S100A2 Human S100A7 Human S100A8 Mouse S100A6 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction