Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

S100A9 Antibody (CPT/1028), Novus Biologicals™
SDP

Mouse Monoclonal Antibody

$280.00 - $524.00

Specifications

Antigen S100A9
Clone CPT/1028
Concentration 0.2 mg/mL
Dilution Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, Immunohistochemistry-Paraffin 0.5 - 1 ug/ml, Immunohistochemistry-Frozen (Negative), Immunofluorescence 0.5 - 1.0 ug/ml
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunofluorescence
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP24491200
SDP
View Documents
Novus Biologicals
NBP2449120.02MG
0.02 mg
Each for $280.00
Only null left
Add to Cart
 
NBP24491201
SDP
View Documents
Novus Biologicals
NBP2449120.1MG
0.1 mg
Each for $524.00
Only null left
Add to Cart
 
NBP24491202
SDP
View Documents
Novus Biologicals
NBP2449120.2MG
0.2 mg N/A N/A N/A
Description

Description

Ensure accurate, reproducible results in Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence, Immunohistochemistry (Frozen) (Negative)

S100A9 Monoclonal specifically detects S100A9 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

S100A9
0.2 mg/mL
Flow Cytometry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunofluorescence
Unconjugated
Mouse
Cancer, Immunology, Innate Immunity, Signal Transduction, Tyrosine Kinases
1.0mM PBS and 0.05% BSA with 0.05% Sodium Azide
6280
Recombinant human Calprotectin protein
Primary
Store at 4C.
14 kDa
CPT/1028
Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, Immunohistochemistry-Paraffin 0.5 - 1 ug/ml, Immunohistochemistry-Frozen (Negative), Immunofluorescence 0.5 - 1.0 ug/ml
Monoclonal
Purified
RUO
Human
CAGBMigration inhibitory factor-related protein 14, calgranulin B, calgranulin-B, Calprotectin L1H subunit, CFAGMRP-14,60B8AG, CGLB, L1AG, LIAG, MAC387, MIF, MRP14Leukocyte L1 complex heavy chain, NIF, P14, protein S100-A9, S100 calcium binding protein A9, S100 calcium binding protein A9 (calgranulin B), S100 calcium-binding protein A9, S100 calcium-binding protein A9 (calgranulin B)
S100A9
IgM κ
Protein A or G purified
Recognizes the L1 or Calprotectin molecule, an intra-cytoplasmic antigen comprising of a 12kDa alpha chain and a 14kDa beta chain. Calprotectin comprises 60% of the cytoplasmic protein fraction of circulating polymorphonuclear granulocytes and is also found in monocytes, macrophages and ileal tissue eosinophils. Peripheral blood monocytes carry the antigen extra- and intracellularly, neutrophils only intracellularly. Calprotectin has antibacterial, antifungal, immunomodulating and antiproliferative effects. Besides this it is a potent chemotactic factor for neutrophils. Plasma concentrations are elevated in diseases associated with increased neutrophil activity, like inflammatory bowel disease. Granulocytes terminate their existence after transmigration through the intestinal wall. Therefore calprotectin is also detectable in feces. Elevated levels of calprotectin have been observed in body fluids such as plasma, saliva, gingival crevicular fluid, stools, and synovial fluid during infection and inflammatory conditions.This MAb reacts with neutrophils, monocytes, macrophages, and squamous mucosal epithelia and is important for identifying macrophages in tissue sections.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.