Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
S100A9 Monoclonal Antibody (OTI13E12), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA804481

Description
MRP-14 is a member of the S100 family of proteins containing 2 EF-hand calcium-binding motifs. S100 proteins are localized in the cytoplasm and/or nucleus of a wide range of cells, and involved in the regulation of a number of cellular processes such as cell cycle progression and differentiation. S100 genes include at least 13 members which are located as a cluster on chromosome 1q21. This protein may function in the inhibition of casein kinase and altered expression of this protein is associated with the disease cystic fibrosis.Specifications
S100A9 | |
Monoclonal | |
Unconjugated | |
S100A9 | |
60B8AG, CAGB, CFAG, CGLB, L1AG, LIAG, MAC387, MIF, MRP14, NIF, P14 | |
Mouse | |
Affinity Chromatography | |
RUO | |
6280 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
OTI13E12 | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
P06702 | |
S100A9 | |
Full length human recombit protein of human S100A9 produced in SF9 cell. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG2a |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction