Learn More
S100A9 Mouse anti-Human, Clone: OTI2A2, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF804217

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
MRP-14 is a member of the S100 family of proteins containing 2 EF-hand calcium-binding motifs. S100 proteins are localized in the cytoplasm and/or nucleus of a wide range of cells, and involved in the regulation of a number of cellular processes such as cell cycle progression and differentiation. S100 genes include at least 13 members which are located as a cluster on chromosome 1q21. This protein may function in the inhibition of casein kinase and altered expression of this protein is associated with the disease cystic fibrosis.Specifications
S100A9 | |
Monoclonal | |
Unconjugated | |
S100A9 | |
60B8AG, CAGB, CFAG, CGLB, L1AG, LIAG, MAC387, MIF, MRP14, NIF, P14 | |
Mouse | |
Affinity Chromatography | |
RUO | |
6280 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Paraffin) | |
OTI2A2 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P06702 | |
S100A9 | |
Full length human recombit protein of human S100A9 produced in SF9 cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.