Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ S100A9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA146489
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA146489 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA146489 Supplier Invitrogen™ Supplier No. PA146489
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Suggested positive control: antigen standard for S100A9 (transient overexpression lysate), HL-60 + DMSO.

S100A9 (MRP-14) is a member of the S100 family of proteins containing 2 EF-hand calcium-binding motifs. S100 proteins are localized in the cytoplasm and/or nucleus of a wide range of cells, and involved in the regulation of a number of cellular processes such as cell cycle progression and differentiation. S100 genes include at least 13 members which are located as a cluster on chromosome 1q21. This protein may function in the inhibition of casein kinase and altered expression of this protein is associated with the disease cystic fibrosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen S100A9
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation Whole serum with 0.1% sodium azide
Gene S100A9
Gene Accession No. P06702, P31725, P50116
Gene Alias 60B8AG; AW546964; BEE22; BM-009; Cagb; calgranulin B; calgranulin-B; Calprotectin L1H subunit; CFAG; CGLB; DKFZp686B04128; GAGB; intracellular calcium-binding protein (MRP14); L1AG; Leukocyte L1 complex heavy chain; LIAG; MAC387; Macrophage; MIF; Migration inhibitory factor-related protein 14; Monocyte; MRP14; MRP-14; MRP15; MRP16; myeloid-related protein 14; NIF; P14; protein S100-A9; S100 A9; S100 calcium binding protein A9; S100 calcium binding protein A9 (calgranulin B); S100 calcium-binding protein A9; S100 calcium-binding protein A9 (calgranulin B); S100a9
Gene Symbols S100A9
Host Species Rabbit
Immunogen Full length human S100A9 protein expressed in E. coli.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20202, 6280, 94195
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.