Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
S100B Antibody (15F4NB) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$158.00 - $501.50
Specifications
Antigen | S100B |
---|---|
Clone | 15F4NB |
Concentration | 1.0 mg/mL |
Dilution | Western Blot 2 μg/mL, Flow Cytometry 1:500, Immunohistochemistry 1:100 - 1:500, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:100 - 1:500, CyTOF-ready |
Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), CyTOF |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP245225SS
![]() |
Novus Biologicals
NBP245225SS |
0.025 mg |
Each for $158.00
|
|
|||||
NBP245225
![]() |
Novus Biologicals
NBP245225 |
0.1 mg |
Each for $501.50
|
|
|||||
Description
Ensure accurate, reproducible results in Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin)
S100B Monoclonal specifically detects S100B in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, CyTOF-ready.Specifications
S100B | |
1.0 mg/mL | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), CyTOF | |
Unconjugated | |
Mouse | |
Alzheimers Research, Apoptosis, Biologically Active Proteins, Breast Cancer, Cancer, Cell Biology, Cellular Markers, Epigenetics, Hypoxia, Lipid and Metabolism, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Peroxisome Markers, Signal Transduction, Stem Cells | |
PBS with No Preservative | |
6285 | |
Partial recombinant human S100B protein (amino acids 2-92) [UniProt P04271] | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
15F4NB | |
Western Blot 2 μg/mL, Flow Cytometry 1:500, Immunohistochemistry 1:100 - 1:500, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:100 - 1:500, CyTOF-ready | |
Monoclonal | |
Purified | |
RUO | |
Human | |
beta (neural), NEF, S100, S100 beta, S100 calcium binding protein B, S100 calcium-binding protein B, S100 calcium-binding protein, beta (neural), S-100 calcium-binding protein, beta chain, 10protein S100-B, S-100 protein beta chain, S-100 protein subunit beta, S100beta | |
S100B | |
IgG2b κ | |
Protein G purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title