Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
S100B Antibody (S1-61), Alexa Fluor™ 594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250282AF594
Description
S100B Monoclonal specifically detects S100B in Human samples. It is validated for Immunohistochemistry.Specifications
S100B | |
Monoclonal | |
Alexa Fluor 594 | |
50 mM sodium borate with 0.05% sodium azide | |
S100B | |
S100B protein conjugated to methylated BSA | |
0.1 mL | |
Primary | |
Human | |
IgG1 |
Immunohistochemistry | |
S1-61 | |
Immunohistochemistry | |
beta (neural), NEF, S100, S100 beta, S100 calcium binding protein B, S100 calcium-binding protein B, S100 calcium-binding protein, beta (neural), S-100 calcium-binding protein, beta chain, 10protein S100-B, S-100 protein beta chain, S-100 protein subunit beta, S100beta | |
Mouse | |
Protein A purified | |
Alzheimers Research, Apoptosis, Biologically Active Proteins, Breast Cancer, Cancer, Cell Biology, Cellular Markers, Epigenetics, Hypoxia, Lipid and Metabolism, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Peroxisome Markers, Signal Transduction, Stem Cells | |
6285 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction