Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ S100B Monoclonal Antibody (CL2720)
GREENER_CHOICE

Catalog No. PIMA534997
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA534997 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA534997 Supplier Invitrogen™ Supplier No. MA534997
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Immunogen sequence: LEKAMVALID VFHQYSGREG DKHKLKKSEL KELINNELSH FLEEIKEQEV VDKVMETLDN DGDGECDFQE FMAFVAMVTT ACHEFFEHE Highest antigen sequence indentity to the following orthologs: Mouse - 99%, Rat - 98%.

S100B is a member of the S100 family of proteins containing 2 EF-hand calcium-binding motifs. S100 proteins are localized in the cytoplasm and/or nucleus of a wide range of cells and are involved in the regulation of a number of cellular processes such as cell cycle progression and differentiation. S100B may function in neurite extension, proliferation of melanoma cells, stimulation of Ca2+ fluxes, inhibition of PKC-mediated phosphorylation, astrocytosis and axonal proliferation, and inhibition of microtubule assembly. Chromosomal rearrangements and altered expression of this gene have been implicated in several diseases, including Alzheimer's disease, Down's syndrome, epilepsy, amyotrophic lateral sclerosis, melanoma, and type I diabetes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen S100B
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone CL2720
Concentration 1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene S100B
Gene Accession No. P04271, P04631, P50114
Gene Alias AI850290; Bpb; hypothetical protein LOC525716; NEF; protein S100-B; S100; S100 calcium binding protein B; S100 calcium binding protein beta (neural); S100 calcium-binding protein B; S100 calcium-binding protein beta (neural); S-100 calcium-binding protein beta subunit; S100 calcium-binding protein, beta (neural); S-100 calcium-binding protein, beta chain; S100 protein beta chain; S-100 protein beta chain; S-100 protein subunit beta; S100 protein, beta polypeptide; S100 protein, beta polypeptide, neural; S100B; S100-B; S100beta; S100P
Gene Symbols S100B
Host Species Mouse
Immunogen Recombinant protein fragment of Human S100B (Product #RP-91304)
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20203, 25742, 6285
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.