Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
S100P Antibody (207), PE

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290136PE
Description
S100P Monoclonal specifically detects S100P in Human samples. It is validated for Flow Cytometry.Specifications
S100P | |
Monoclonal | |
PE | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
207 | |
Flow Cytometry | |
MIG9, migration-inducing gene 9, Protein S100-E, S100 calcium binding protein P, S100 calcium-binding protein Pprotein S100-P, S100E | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human S100P (P25815; Met1-Lys95). | |
0.1 mL | |
Cancer | |
6286 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction