Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
S100P Antibody (OTI1A4), DyLight 755, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273972IR
Description
S100P Monoclonal specifically detects S100P in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
S100P | |
Monoclonal | |
DyLight 755 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
S100P | |
Full length recombinant protein of human S100P (NP_005971) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human | |
IgG1 |
Western Blot, Immunocytochemistry, Immunofluorescence | |
OTI1A4 | |
Western Blot, Immunocytochemistry/Immunofluorescence | |
MIG9, migration-inducing gene 9, Protein S100-E, S100 calcium binding protein P, S100 calcium-binding protein Pprotein S100-P, S100E | |
Mouse | |
Affinity Purified | |
Cancer | |
6286.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction