Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ S100P Monoclonal Antibody (357517)
GREENER_CHOICE

Catalog No. PIMA524106
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA524106 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA524106 Supplier Invitrogen™ Supplier No. MA524106
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) S100A10 or rhS100B is observed. Reconstitute at 0.5 mg/mL in sterile PBS.

The protein encoded by this gene is a member of the S100 family of proteins containing 2 EF-hand calcium-binding motifs. S100 proteins are localized in the cytoplasm and/or nucleus of a wide range of cells, and involved in the regulation of a number of cellular processes such as cell cycle progression and differentiation. S100 genes include at least 13 members which are located as a cluster on chromosome 1q21; however, this gene is located at 4p16. This protein, in addition to binding Ca2+, also binds Zn2+ and Mg2+. This protein may play a role in the etiology of prostate cancer.
TRUSTED_SUSTAINABILITY

Specifications

Antigen S100P
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 357517
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene S100P
Gene Accession No. P25815
Gene Alias MIG9; Migration-inducing gene 9 protein; protein S100-E; Protein S100-P; S100 calcium binding protein P; S100 calcium-binding protein P; S100E; S100p
Gene Symbols S100P
Host Species Mouse
Immunogen E. coli-derived recombinant human S100P Met1-Lys95.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6286
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.