Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
S100P Monoclonal Antibody (OTI1A4), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA504478S


Description
S100P Monoclonal antibody specifically detects S100P in Human samples. It is validated for Immunocytochemistry, Western BlotSpecifications
S100P | |
Monoclonal | |
1.84 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
P25815 | |
Mouse | |
Affinity chromatography | |
RUO | |
6286 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG1 |
Immunocytochemistry, Western Blot | |
OTI1A4 | |
Unconjugated | |
S100P | |
MIG9; Migration-inducing gene 9 protein; protein S100-E; Protein S100-P; S100 calcium binding protein P; S100 calcium-binding protein P; S100E; S100p | |
Full length human recombinant protein of human S100P produced in HEK293T cell. | |
30 μL | |
Primary | |
Human | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction