Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ S100PBP Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA544508
Description
Peptide sequence: ERRLGKVIPV LQTKTRTNVP TFSQSNLEQQ KQLYLRSVIA HIEDPEDTNQ Sequence homology: Cow: 85%; Dog: 85%; Horse: 86%; Human: 100%; Mouse: 85%; Pig: 85%; Rabbit: 77%; Rat: 85%.
The specific function of this protein remains unknown.
Specifications
| S100PBP | |
| Polyclonal | |
| Unconjugated | |
| S100PBP | |
| 4930429A08Rik; AI851343; hypothetical protein LOC539983; RGD1564943; S100P binding protein; S100P binding protein 1; S100P-binding protein; S100P-binding protein Riken; S100pbp; S100PBPR | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 64766 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q96BU1 | |
| S100PBP | |
| Synthetic peptide directed towards the middle region of human S100PBP. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction