Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ S100PBP Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA555555
Description
Immunogen sequence: SRVPSEQSSG TSLLPKDGAP FSWDSLDEDG LDDSLLELSE GEEDDGDVNY TEEEIDALLK EDDPSYEQSS GEDDGGHVEK GERGSQILLD TP Highest antigen sequence identity to the following orthologs: Mouse - 60%, Rat - 62%.
The specific function of this protein remains unknown.
Specifications
S100PBP | |
Polyclonal | |
Unconjugated | |
S100PBP | |
4930429A08Rik; AI851343; hypothetical protein LOC539983; RGD1564943; S100P binding protein; S100P binding protein 1; S100P-binding protein; S100P-binding protein Riken; S100pbp; S100PBPR | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
64766 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96BU1 | |
S100PBP | |
Recombinant protein corresponding to Human S100PBP. Recombinant protein control fragment (Product #RP-94080). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction