Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC13A4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110376
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110376 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5110376 Supplier Invitrogen™ Supplier No. PA5110376
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: WFTREPGFVP GWDSFFEKKG YRTDATVSVF LGFLLFLIPA KKPCFGKKND GENQEHSLGT EPIITWKDFQ KTMPWE.

SLC13A4 gene ontology annotations related to this gene include sodium ion transport; transmembrane transport.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC13A4
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.23 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Slc13a4
Gene Accession No. Q9UKG4
Gene Alias 9630060C05Rik; Na(+)/sulfate cotransporter SUT-1; NAS2; SLC13A4; sodium sulfate cotransporter-2; solute carrier family 13 (sodium/sulfate symporter), member 4; solute carrier family 13 (sodium/sulfate symporters), member 4; solute carrier family 13 (sodium/sulphate symporters), member 4; solute carrier family 13 member 4; sulphate transporter 1; SUT1; SUT-1
Gene Symbols Slc13a4
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 390-465 of human SLC13A4 (NP_036582.2).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 243755, 26266, 503568
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.