Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
S1P1/EDG-1 Antibody (218713), DyLight 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB2016K
Description
S1P1/EDG-1 Monoclonal antibody specifically detects S1P1/EDG-1 in Human samples. It is validated for Flow Cytometry, CyTOFSpecifications
S1P1/EDG-1 | |
Monoclonal | |
DyLight 488 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry, CyTOF | |
218713 | |
Flow Cytometry, CyTOF-ready | |
CD363, CD363 antigen, CHEDG1, D1S3362, EDG-1, EDG1edg-1, Endothelial differentiation G-protein coupled receptor 1, endothelial differentiation, sphingolipid G-protein-coupled receptor, 1, FLJ58121, S1P receptor 1, S1P receptor Edg-1, s1p1, S1P1ECGF1, sphingosine 1-phosphate receptor 1, sphingosine 1-phosphate receptor EDG1, Sphingosine 1-phosphate receptor Edg-1, sphingosine-1-phosphate receptor 1 | |
NS0 mouse myeloma cell line transfected with human S1P1/EDG-1, Met1-Ser382, Accession # P21453 | |
0.1 mL | |
Cancer, Cell Biology, Cellular Markers, GPCR, Lipid and Metabolism, Neuroscience, Signal Transduction | |
1901 | |
Store at 4°C in the dark. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction