Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
S1P1/EDG-1 Antibody (713412) [PerCP], Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB7089C
Description
S1P1/EDG-1 Monoclonal antibody specifically detects S1P1/EDG-1 in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, CyTOFSpecifications
S1P1/EDG-1 | |
Monoclonal | |
PerCP | |
PBS | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Mouse | |
Purified |
Flow Cytometry, Immunohistochemistry, CyTOF | |
713412 | |
Flow Cytometry, Immunohistochemistry, CyTOF-ready | |
CD363, CD363 antigen, CHEDG1, D1S3362, EDG-1, EDG1edg-1, Endothelial differentiation G-protein coupled receptor 1, endothelial differentiation, sphingolipid G-protein-coupled receptor, 1, FLJ58121, S1P receptor 1, S1P receptor Edg-1, s1p1, S1P1ECGF1, sphingosine 1-phosphate receptor 1, sphingosine 1-phosphate receptor EDG1, Sphingosine 1-phosphate receptor Edg-1, sphingosine-1-phosphate receptor 1 | |
Mouse S1P1/EDG-1 synthetic peptide (T4-H28), Accession # O08530 | |
0.1 mL | |
Cancer, Cell Biology, Cellular Markers, GPCR, Lipid and Metabolism, Neuroscience, Signal Transduction | |
1901 | |
Store at 4°C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction