Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
S1P1/EDG-1 Antibody (MM0045-21L9), DyLight 755, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB11093513IR
Description
S1P1/EDG-1 Monoclonal specifically detects S1P1/EDG-1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
S1P1/EDG-1 | |
Monoclonal | |
DyLight 755 | |
P21453 | |
S1PR1 | |
Human recombinant S1P1/EDG-1 | |
0.1 mL | |
Cancer, Cytoskeleton Markers, GPCR, Lipid and Metabolism, Neuroscience, Signal Transduction | |
1901 | |
Human | |
Purified |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
MM0045-21L9 | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin | |
CD363, CD363 antigen, CHEDG1, D1S3362, EDG-1, EDG1edg-1, Endothelial differentiation G-protein coupled receptor 1, endothelial differentiation, sphingolipid G-protein-coupled receptor, 1, FLJ58121, S1P receptor 1, S1P receptor Edg-1, s1p1, S1P1ECGF1, sphingosine 1-phosphate receptor 1, sphingosine 1-phosphate receptor EDG1, Sphingosine 1-phosphate receptor Edg-1, sphingosine-1-phosphate receptor 1 | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
This recognizes human EDG-1 in western blot. | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction