Learn More
Invitrogen™ S1P1 Monoclonal Antibody (2B9)

Description
Mouse anti Human Sphingosine 1-Phosphate Receptor 1, clone 2B9 recognizes S1P1, otherwise known as CD363/EDG-1, a bioactive lysophospholipid and inducible member of the G-protein coupled receptor superfamily (GPCR) which plays an important role in a variety of cellular processes.
Specifications
Specifications
| Antigen | S1P1 |
| Applications | Flow Cytometry, Immunoprecipitation, Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 2B9 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with <0.1% sodium azide |
| Gene | S1PR1 |
| Gene Accession No. | P21453 |
| Gene Alias | AI849002; CD antigen CD363; CD363; CD363 antigen; CHEDG1; D1S3362; ECGF1; EDG1; EDG-1; EDG1 (Edg1); Edg1; rhodopsin family G protein-coupled receptor; endothelial differentiation G-protein coupled receptor 1; endothelial differentiation sphingolipid G-protein-coupled receptor 1; endothelial differentiation, sphingolipid G-protein-coupled receptor, 1; etID43307.22; Lpb1; Lysophospholipid receptor B1; S1p; S1P receptor 1; S1P receptor Edg-1; s1p1; S1PR1; sphingosine 1-phosphate receptor; sphingosine 1-phosphate receptor 1; sphingosine 1-phosphate receptor EDG1; Sphingosine 1-phosphate receptor Edg-1; sphingosine-1-phosphate receptor 1; wu:fb98f05 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.