Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ S1P1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. OPA115004
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OPA115004 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. OPA115004 Supplier Invitrogen™ Supplier No. OPA115004
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Control: Heart, Vessel.

S1P1 is a G-protein coupled receptor for the bioactive lysosphingolipid sphingosine 1-phosphate (S1P) that is coupled to the G(i) subclass of heteromeric G proteins. S1P1 signaling leads to the activation of RAC1, SRC, PTK2/FAK1 and MAP kinases. It plays an important role in cell migration via its role in the reorganization of the actin cytoskeleton and formation of lamellipodia in response to stimuli that increase the activity of the sphingosine kinase SPHK1. S1P1 is required for normal chemotaxis toward sphingosine 1-phosphate, normal embryonic heart development, and normal cardiac morphogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen S1P1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene S1PR1
Gene Accession No. P21453
Gene Alias AI849002; CD antigen CD363; CD363; CD363 antigen; CHEDG1; D1S3362; ECGF1; EDG1; EDG-1; EDG1 (Edg1); Edg1; rhodopsin family G protein-coupled receptor; endothelial differentiation G-protein coupled receptor 1; endothelial differentiation sphingolipid G-protein-coupled receptor 1; endothelial differentiation, sphingolipid G-protein-coupled receptor, 1; etID43307.22; Lpb1; Lysophospholipid receptor B1; S1p; S1P receptor 1; S1P receptor Edg-1; s1p1; S1PR1; sphingosine 1-phosphate receptor; sphingosine 1-phosphate receptor 1; sphingosine 1-phosphate receptor EDG1; Sphingosine 1-phosphate receptor Edg-1; sphingosine-1-phosphate receptor 1; wu:fb98f05
Gene Symbols S1PR1
Host Species Rabbit
Immunogen Synthetic 16 amino acid peptide from 1st cytoplasmic domain of human S1PR1/EDG1.
Purification Method Antigen affinity chromatography
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1901
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.