Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
S1P3/EDG-3 Rabbit anti-Human, Mouse, Rat, Clone: 5E3G9, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $494.50
Specifications
Antigen | S1P3/EDG-3 |
---|---|
Clone | 5E3G9 |
Applications | Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
S1P3/EDG-3 Monoclonal antibody specifically detects S1P3/EDG-3 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
S1P3/EDG-3 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Cell Biology, Cell Cycle and Replication, Cellular Markers, GPCR, Lipid and Metabolism, Signal Transduction | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
1903 | |
IgG | |
Affinity purified |
5E3G9 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
EDG-3, EDG3FLJ37523, Endothelial differentiation G-protein coupled receptor 3, endothelial differentiation, sphingolipid G-protein-coupled receptor, 3, FLJ93220, G protein-coupled receptor, endothelial differentiation gene-3, LPB3, MGC71696, S1P receptor 3, S1P receptor Edg-3, S1P receptor EDG3, S1P3, sphingosine 1-phosphate receptor 3, Sphingosine 1-phosphate receptor Edg-3, sphingosine-1-phosphate receptor 3 | |
A synthetic peptide corresponding to a sequence within amino acids 1-100 of human S1P3/EDG-3 (Q99500). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title