Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
S1P4/EDG-6 Antibody, PE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP224500PE
Description
S1P4/EDG-6 Polyclonal antibody specifically detects S1P4/EDG-6 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
S1P4/EDG-6 | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin | |
EDG6S1P receptor Edg-6, Endothelial differentiation G-protein coupled receptor 6, endothelial differentiation, G-protein-coupled receptor 6, endothelial differentiation, lysophosphatidic acid G-protein-coupled receptor6,endothelial differentiation, G protein coupled receptor 6, S1P receptor 4, S1P4LPC1, SLP4, sphingosine 1-phosphate receptor 4, Sphingosine 1-phosphate receptor Edg-6, sphingosine-1-phosphate receptor 4 | |
A portion of amino acids 1-50 of human Sphingolipid Receptor Edg6/S1P4 was used as the immunogen. | |
0.1 mL | |
Cell Biology, Cellular Markers, GPCR, Signal Transduction | |
8698 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
PE | |
PBS | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction