Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
S1P5/EDG-8 Antibody (1196A), mFluor Violet 450 SE, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals FAB9084MFV450
Description
S1P5/EDG-8 Monoclonal antibody specifically detects S1P5/EDG-8 in Mouse samples. It is validated for Western Blot, Flow CytometrySpecifications
| S1P5/EDG-8 | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| 0.1 mL | |
| GPCR | |
| 53637 | |
| Store at 4C in the dark. |
| Western Blot, Flow Cytometry | |
| 1196A | |
| Western Blot, Flow Cytometry | |
| Edg-8, EDG8S1P receptor Edg-8, Endothelial differentiation G-protein-coupled receptor 8, endothelial differentiation, sphingolipid G-protein-coupled receptor, 8, S1P receptor 5, S1P5, sphingosine 1-phosphate receptor 5, sphingosine 1-phosphate receptor EDG8, Sphingosine 1-phosphate receptor Edg-8, sphingosine-1-phosphate receptor 5, SPPR-1, SPPR-2 | |
| Mouse S1P5/EDG-8 peptide corresponding to the N-terminal extracellular sequence, Accession # Q91X56 | |
| RUO | |
| Primary | |
| Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction