Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
S1P5/EDG-8 Antibody (282503), Janelia Fluor™ 646, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB3964J
Description
S1P5/EDG-8 Monoclonal antibody specifically detects S1P5/EDG-8 in Human samples. It is validated for Flow Cytometry, CyTOFSpecifications
S1P5/EDG-8 | |
Monoclonal | |
Janelia Fluor 646 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry, CyTOF | |
282503 | |
Flow Cytometry, CyTOF-ready | |
Edg-8, EDG8S1P receptor Edg-8, Endothelial differentiation G-protein-coupled receptor 8, endothelial differentiation, sphingolipid G-protein-coupled receptor, 8, S1P receptor 5, S1P5, sphingosine 1-phosphate receptor 5, sphingosine 1-phosphate receptor EDG8, Sphingosine 1-phosphate receptor Edg-8, sphingosine-1-phosphate receptor 5, SPPR-1, SPPR-2 | |
BaF3 mouse pro-B cell line transfected with human S1P5/EDG-8, Met1-Asp398, Accession # Q9H228 | |
0.1 mL | |
GPCR | |
53637 | |
Store at 4°C in the dark. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction