Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ S1PR2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA572265
Description
S1PR2 Polyclonal Antibody for Western Blot
This gene encodes a member of the G protein-coupled receptors, as well as the EDG family of proteins. The encoded protein is a receptor for sphingosine 1-phosphate, which participates in cell proliferation, survival, and transcriptional activation. Defects in this gene have been associated with congenital profound deafness.
Specifications
S1PR2 | |
Polyclonal | |
Unconjugated | |
S1PR2 | |
agr16; edg5; edg-5; S1PR2 | |
Rabbit | |
Antigen Affinity Chromatography, Protein A | |
RUO | |
170457 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.44 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
Q9I8K8 | |
S1PR2 | |
KLH conjugated synthetic peptide between 39-73 amino acids from DANRE s1pr2. | |
200 μL | |
Primary | |
Zebrafish | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction