Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
S5a/Angiocidin Antibody (CPTC-PSMD4-3), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308271B
Description
S5a/Angiocidin Monoclonal antibody specifically detects S5a/Angiocidin in Human samples. It is validated for Western Blot, Protein ArraySpecifications
S5a/Angiocidin | |
Monoclonal | |
Biotin | |
26S proteasome non-ATPase regulatory subunit 4;AF;AF-1;ASF;MCB1;PSMD4;pUB-R5;Rpn10;S5A | |
Recombinant full-length human S5a/Angiocidin protein (Uniprot: P55036) | |
100 μg | |
Neuroscience, Ubiquitin Proteasome Pathway | |
5710 | |
Store at 4°C in the dark. | |
IgG2b κ |
Western Blot, Protein Array | |
CPTC-PSMD4-3 | |
PBS | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction