Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
S5a/Angiocidin Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB1203318
Description
S5a/Angiocidin Polyclonal specifically detects S5a/Angiocidin in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
Proteasome 19S S5A | |
Polyclonal | |
Unconjugated | |
PBS containing 1 mg/ml BSA with 0.05% Sodium Azide | |
26S proteasome non-ATPase regulatory subunit 4, 26S proteasome regulatory subunit S5A, AF-1,26S protease subunit S5a, AFMCB1, angiocidin, Antisecretory factor 1, ASF, multiubiquitin chain binding protein, Multiubiquitin chain-binding protein, proteasome (prosome, macropain) 26S subunit, non-ATPase, 4, pUB-R5,26S proteasome regulatory subunit RPN10, Rpn10, RPN10 homolog, S5A, S5a/antisecretory factor protein | |
Rabbit | |
Ammonium sulfate precipitation | |
RUO | |
Primary | |
Detects proteasome 19S subunit S5A. | |
Store at -20C. Avoid freeze-thaw cycles. | |
IgG |
Western Blot, Immunoprecipitation | |
1.0mg/mL | |
Western Blot 1:500, Immunoprecipitation 1:10 - 1:500 | |
P55036 | |
PSMD4 | |
Human recombinant proteasome 19S subunit S5A. | |
200 μL | |
Neuroscience, Ubiquitin Proteasome Pathway | |
5710 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction