Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ S7A6O Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559614
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559614 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559614 Supplier Invitrogen™ Supplier No. PA559614
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AEALVLACKR LRSDAVESAA QKTSEGLERA AENNVFHLVA TVCSQEEPVQ PLLREVLRPS RDSQQRVRRN LRASAREVRQ EGRYRVLS Highest antigen sequence identity to the following orthologs: Mouse - 73%, Rat - 73%.

TRUSTED_SUSTAINABILITY

Specifications

Antigen S7A6O
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SLC7A6OS
Gene Accession No. Q96CW6
Gene Alias ADAMS proteinase-related protein; Probable RNA polymerase II nuclear localization protein SLC7A6OS; protein SLC7A6OS; SLC7A6OS; solute carrier family 7 member 6 opposite strand; solute carrier family 7 member 6 opposite strand transcript; solute carrier family 7, member 6 opposite strand
Gene Symbols SLC7A6OS
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human S7A6O. Recombinant protein control fragment (Product #RP-98542).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84138
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.