Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ SA 4503 dihydrochloride
GREENER_CHOICE

Catalog No. 495110
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
49-511-0 10 mg
49-515-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 49-511-0 Supplier Tocris Bioscience™ Supplier No. 4951/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Selective σ1 receptor agonist

Selective σ1 receptor agonist (IC50 = 17.4 nM). Exhibits selectivity for σ1 over σ2 receptors. Inhibits angiotensin II-induced cardiomyocyte hypertrophy in vitro and attenuates myocardial hypertrophy in vivo. Enhances brain plasticity and sensorimotor function in a rat model of experimental stroke.

Chemical Identifiers

CAS 165377-44-6
Molecular Formula C23H34Cl2N2O2

Specifications

Quantity 10 mg
Formula Weight 441.43
Physical Form Solid
Color White
Purity >98%
Molecular Weight (g/mol) 441.437
Chemical Name or Material SA 4503 dihydrochloride
Recommended Storage Desiccate at RT
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.