Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SAS™ StrepAlert™ (Dipstick) Strep A Test Kit

Catalog No. sacol611530
Change view
Click to view available options
Quantity:
30 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SACOL611530 30 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. SACOL611530 Supplier SAS™ Supplier No. COL611530
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Includes

Test device strips, throat swabs, extraction tubes, reagents A and B, positive and negative controls, and package insert

Membrane immunoassay kit with results in as little as five minutes

  • CLIA Complexity: Waived and Non-waived
  • CPT Codes: 87880QW (Waived); 87880 (Non-waived)
  • Sensitivity: 97.9%
  • Specificity: 97.3%
  • Positive and negative controls

Specifications

CPT Code 87880, 87880QW
Clia Complexity Waived, Non-waived
Detectable Analytes Streptococcal Antigen (Group A)
Type Streptococcus A Test Kit
Includes Test device strips, throat swabs, extraction tubes, reagents A and B, positive and negative control and package insert
Quantity 30 Tests
Test Time 5 min.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.