Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SA1 Antibody, Alexa Fluor™ 488, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NB100298AF488
Description
SA1 Polyclonal antibody specifically detects SA1 in Human, Mouse, Invertebrate samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
SA1 | |
Polyclonal | |
Western Blot, Immunocytochemistry/Immunofluorescence | |
cohesin subunit SA-1, nuclear protein stromal antigen 1, SA1, stromal antigen 1 | |
A 36 amino acid synthetic peptide, which represented a portion of human Stromal Antigen 1 encoded in part by exons 5 and 6 (LocusLink ID 10274). | |
0.1 mL | |
Cell Cycle and Replication, DNA Repair, Mitotic Regulators | |
10274 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunofluorescence | |
Alexa Fluor 488 | |
50mM Sodium Borate | |
Goat | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse, Invertebrate | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction