Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SA1 Antibody, Novus Biologicals™

Goat Polyclonal Antibody has been used in 2 publications
Supplier: Novus Biologicals NB100298
Description
SA1 Polyclonal specifically detects SA1 in Human, Mouse, Invertebrate samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.Specifications
SA1 | |
Polyclonal | |
Unconjugated | |
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
STAG1 | |
A 36 amino acid synthetic peptide, which represented a portion of human Stromal Antigen 1 encoded in part by exons 5 and 6 (LocusLink ID 10274). | |
100 μL | |
Cell Cycle and Replication, DNA Repair, Mitotic Regulators | |
10274 | |
Human, Mouse, Invertebrate | |
IgG |
Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
1.0 mg/mL | |
Western Blot 1:1000-1:10000, Immunocytochemistry/Immunofluorescence 1:500-1:2000, Immunoprecipitation | |
cohesin subunit SA-1, nuclear protein stromal antigen 1, SA1, stromal antigen 1 | |
Goat | |
Affinity Purified | |
RUO | |
Primary | |
NB100-298 is specific for human SA1 protein. Human, Xenopus. Based on 100% sequence identity, this antibody is predicted to react with Mouse, Rhesus Monkey, Crab-eating macaque and Northern white-cheeked gibbon. | |
Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction