Learn More
SA2 Polyclonal Antibody, Bethyl Laboratories
Goat Polyclonal Antibody
$151.31
Specifications
Antigen | SA2 |
---|---|
Applications | Immunocytochemistry, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-155-7006
![]() |
Bethyl Laboratories, Inc
A300158AT |
20 μL |
Each of 1 for $151.31
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse
STAG2 is a component of the cohesin complex. This complex is required for the cohesion of sister chromatids after DNA replication. The cohesin complex forms a large proteinaceous ring within which sister chromatids can be trapped. At anaphase, the complex is cleaved and dissociates from chromatin, allowing sister chromatids to segregate. The cohesin complex may also play a role in spindle pole assembly during mitosis.Specifications
SA2 | |
Polyclonal | |
Liquid | |
RUO | |
TBS with 0.1% BSA and 0.09% sodium azide | |
STAG2; stromal antigen 2; RP11-517O1.1; DKFZp686P168; DKFZp781H1753; FLJ25871; SA-2; SA2; bA517O1.1; SCC3 homolog 2 | |
STAG2 | |
IgG | |
Antigen affinity chromatography | |
Antibody |
Immunocytochemistry, Western Blot | |
Unconjugated | |
Goat | |
Human | |
Q8N3U4 | |
10735 | |
Between 1100 and 1150 | |
Primary | |
4° C | |
STAG2 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.