Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SAA1 Rabbit anti-Human, Unconjugated, Clone: ABAE-19, Abnova™

Catalog No. 89337102 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-337-102 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-337-102 Supplier Abnova Corporation Supplier No. MAB20702
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody.

SAA1 Monoclonal antibody specifically detects SAA1 in Human samples. It is validated for IHC, WB-Ti.

Specifications

Antigen SAA1
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone ABAE-19
Conjugate Unconjugated
Description serum amyloid A1
Gene SAA1
Gene Alias MGC111216, PIG4, SAA, TP53I4
Host Species Rabbit
Immunogen A synthetic peptide corresponding to human SAA1
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 6288
Target Species Human
Content And Storage Store at -20°C for one year. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for a longer time. Avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.