Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SAA2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5115862
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5115862 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5115862 Supplier Invitrogen™ Supplier No. PA5115862
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total Serum Amyloid A2.

Major acute phase reactant. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen SAA2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene SAA2
Gene Accession No. P0DJI9
Gene Alias Amyloid fibril protein AA; Amyloid protein A; AW111173; LOC100009259; LOW QUALITY PROTEIN: serum amyloid A-2 protein; SAA; Saa1; Saa2; Saa-2; serum amyloid A 1; serum amyloid A 2; serum amyloid A protein; Serum amyloid A1 protein; serum amyloid A2; Serum amyloid A-2 protein; serum amyloid protein A; unknown; Unknown (protein for MGC:133484)
Gene Symbols SAA2
Host Species Rabbit
Immunogen A synthesized peptide derived from human SAA2(Accession P0DJI9), corresponding to amino acid residues T5-H55.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6289
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.