Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SAA4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA542137
Description
Peptide sequence: AAKLISRSRV YLQGLIDYYL FGNSSTVLED SKSNEKAEEW GRSGKDPDRF Sequence homology: Human: 100%.
Serum Amyloid A4 (SAA4) is a gene that encodes one of the constitutively expressed serum amyloid proteins. Located on chromosome 11, SAA4 is part of the serum amyloid A (SAA) superfamily, which includes other highly homologous genes like SAA1, SAA2, and SAA3. Unlike its counterparts, SAA4 is not an acute-phase reactant but is persistently expressed across different tissues, including plasma, where it associates with high-density lipoprotein (HDL). SAA4 is involved in lipid metabolism and inflammatory responses, albeit not directly induced by inflammatory stimuli like other SAA proteins. It appears as multiple isoforms, some of which may be glycosylated. Recent studies have shown that SAA4 expression encompasses a broad spectrum of physiological conditions and may serve as a marker in diseases involving chronic inflammation and persistent pathological states such as neovascular age-related macular degeneration. The gene's constitutive expression pattern suggests its fundamental role in maintaining homeostasis rather than responding dynamically to acute inflammatory challenges.
Specifications
| SAA4 | |
| Polyclonal | |
| Unconjugated | |
| Saa4 | |
| amyloid A-5 protein; Constitutively expressed serum amyloid A protein; CSAA; C-SAA; Saa4; Saa-4; Saa5; Saa-5; serum amyloid A 4; serum amyloid A-4 protein; serum amyloid A4, constitutive | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 6291 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| P35542 | |
| Saa4 | |
| Synthetic peptide directed towards the middle region of human SAA4. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction