Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SAA4 Rabbit anti-Human, Polyclonal, Proteintech
SDP

Catalog No. 501732868 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-2868 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-2868 Supplier Proteintech Group Inc Supplier No. 203981AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SAA4 is a major acute phase reactant. It is an Apolipoprotein of the HDL complex.

Specifications

Antigen SAA4
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.19 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene SAA4
Gene Accession No. P35542
Gene Alias C SAA, CSAA, SAA4, Serum amyloid A 4 protein, serum amyloid A4, constitutive
Gene Symbols SAA4
Host Species Rabbit
Immunogen SAA4 Fusion Protein Ag14081
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6291
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.