Learn More
Invitrogen™ SAA4 Recombinant Rabbit Monoclonal Antibody (014)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA530833
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire. This antibody has specificity for Human SAA4.
Serum Amyloid A4 (SAA4) is a gene that encodes one of the constitutively expressed serum amyloid proteins. Located on chromosome 11, SAA4 is part of the serum amyloid A (SAA) superfamily, which includes other highly homologous genes like SAA1, SAA2, and SAA3. Unlike its counterparts, SAA4 is not an acute-phase reactant but is persistently expressed across different tissues, including plasma, where it associates with high-density lipoprotein (HDL). SAA4 is involved in lipid metabolism and inflammatory responses, albeit not directly induced by inflammatory stimuli like other SAA proteins. It appears as multiple isoforms, some of which may be glycosylated. Recent studies have shown that SAA4 expression encompasses a broad spectrum of physiological conditions and may serve as a marker in diseases involving chronic inflammation and persistent pathological states such as neovascular age-related macular degeneration. The gene's constitutive expression pattern suggests its fundamental role in maintaining homeostasis rather than responding dynamically to acute inflammatory challenges.
Specifications
SAA4 | |
Recombinant Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
P35542 | |
Saa4 | |
Recombinant Human SAA4 Protein (Glu19-Tyr130). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
ELISA | |
014 | |
Unconjugated | |
Saa4 | |
amyloid A-5 protein; Constitutively expressed serum amyloid A protein; CSAA; C-SAA; Saa4; Saa-4; Saa5; Saa-5; serum amyloid A 4; serum amyloid A-4 protein; serum amyloid A4, constitutive | |
Rabbit | |
Protein A | |
RUO | |
6291 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.