Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SAB Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA529416
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA529416 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA529416 Supplier Invitrogen™ Supplier No. PA529416
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: THP-1. Predicted reactivity: Mouse (91%), Rat (91%), Chicken (81%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Inhibits the auto- and transphosphorylation activity of BTK. Plays a negative regulatory role in BTK-related cytoplasmic signaling in B-cells. May be involved in BCR-induced apoptotic cell death.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SAB
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.65 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene SH3BP5
Gene Accession No. O60239
Gene Alias AI606498; Sab; SH3 binding protein; SH3 domain binding protein 5; SH3 domain-binding protein 5; SH3 domain-binding protein that preferentially associates with BTK; SH3BP5; SH3BP-5; SH3-domain binding protein 5 (BTK-associated); Vascular endothelial cell-specific protein 18; Vesp18
Gene Symbols SH3BP5
Host Species Rabbit
Immunogen Full length human SH3BP5 Recombinant protein.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9467
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.