Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sabinene hydrate, ≥97% (GC), MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco Products
Click to view available options
Quantity:
500 mg
5 g
Description
Featured Industries: Cleaning Products, Cosmetics, Food and Beverages and Personal Care
Specifications
Specifications
| CAS | 546-79-2 |
| Physical Form | Neat |
| Linear Formula | C10H18 O |
| Melting Point | 58°C to 62°C |
| Synonym | 4-Thujanol |
| Type | Analytical Standard |
| Quantity | 500 mg |
| Formula Weight | 154.25 |
| Percent Purity | ≥97% (GC) |
| Grade | Analytical Standard |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction