Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ Sabouraud Dextrose Agar
Click to view available options
Quantity:
10/Pk.
100/Pk.
20/Pk.
Format:
1 oz. Bottle
20 x 150mm Pour Tube (G-PT)
20mm x 150mm Tube - Slant
BB - 200mL
BB - 500mL
BRB - 100mL
Monoplate Deep Pour
Manufacturing Origin:
United States
Description
Isolate, cultivate and differentiate dermatophytes, fungi and yeasts, such as Candida albicans, using acidic pH Thermo Scientific™ Sabouraud Glucose Agar.
Fungi maintain their typical cultural appearance and may thus be readily identified according to the standard macroscopic characteristics. Infections caused by Candida spp. are diverse, ranging from superficial and local to deep-seated and disseminated. Candida albicans and Candida glabrata comprise 70-80% of yeast isolates recovered from patients with invasive disease. This culture media, also available in powder form, has applications in environmental monitoring and sterility testing, enabling corrective measures to be implemented rapidly if unacceptable levels of microbial loading indicate lapses in process control.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.

Specifications
Specifications
Certifications/Compliance | Industrial Reference: BAM, COMPF, USP. |
Color | Amber |
Content And Storage | 2 to 25°C |
Format | 20 x 150mm Pour Tube (G-PT) |
Manufacturing Origin | United States |
Product Type | Agar |
Fill Volume | 20 mL |
Quantity | 100/Pk. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction