Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SAC3D1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA560076
Description
Immunogen sequence: AFREGNAARL FRLLQTLPYL PSCAVQCHVG HARREALARF ARAFSTPKGQ TLPLGFMVNL LALDGLREAR DLCQAHGLPL DGEERVVFLR GRYVEEGLPP ASTCKVLVE Highest antigen sequence identity to the following orthologs: Mouse - 72%, Rat - 72%.

Specifications
SAC3D1 | |
Polyclonal | |
Unconjugated | |
SAC3D1 | |
HSU79266; SAC3 domain containing 1; SAC3 domain-containing protein 1; Sac3 homology domain 1; SAC3 homology domain-containing protein 1; SAC3D1; SHD1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
29901 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.6 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
A6NKF1 | |
SAC3D1 | |
Recombinant protein corresponding to Human SAC3D1. Recombinant protein control fragment (Product #RP-97464). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction