Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SaCas9 Mouse anti-Bacteria, Clone: 11C12, GenScript™

Catalog No. 502120673
Encompass_Preferred
Change view
Click to view available options
Quantity:
40 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-212-0673 40 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-212-0673 Supplier Genscript Corporation Supplier No. A0195140
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reconstitute the lyophilized antibody with deionized water (or equivalent) to a final concentration of 0.5 mg/mL. Clustered regularly interspaced short palindromic repeats (CRISPR)/CRISPR-associated (Cas) protein 9 system provides a robust and multiplexable genome editing tool, enabling researchers to precisely manipulate specific genomic elements, and facilitating the elucidation of target gene function in biology and diseases. CRISPR/Cas9 genome editing allows for double-stranded DNA breaks at specific sequences to efficiently disrupt, excise, mutate, insert, or replace genes. The best characterized CRISPR-associated nucleases are the Cas9 proteins from Streptococcus pyogenes and Staphylococcus aureus. It is important that the precision of transfection and the level of Cas9 expression should be controlled strictly during the editing processes by using specific anti-CRISPR/Cas9 antibodies.

SaCas9 Monoclonal antibody specifically detects SaCas9 in Bacteria samples. It is validated for ELISA, Immunocytochemistry, Immunoprecipitation, Western Blot

Specifications

Antigen SaCas9
Applications ELISA, Immunocytochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 11C12
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide; pH 7.4
Gene Alias cas9; CASP 9; Caspase9; Caspase-9 precursor; CASPASE-9c; CRISPR9; CRISPR-Cas9; CTD; OTTHUMP00000002324; RNCASP9; RP11-265F14.3; s aureus
Host Species Mouse
Immunogen Recombinant Staphylococcus aureus CRISPR/Cas9
Purification Method Protein A
Quantity 40 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Lyophilized
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.