Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Saccharin, Powder, NF, 99-101%, Spectrum™ Chemical
SDP

Catalog No. 18606130 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
Packaging:
Poly Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-606-130 2.5 kg Poly Pail
18-606-128 125 g Poly Bottle
18-606-129 500 g Poly Bottle
18-606-131 12 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-606-130 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. SA1052.5KG
Add to Cart
Add to Cart

SA105, 81-07-2, C7H5NO3S

Spectrum™ Chemical Saccharin, Powder, NF is a pharmaceutical grade substance whose most common use is as an artificial sweetener. All Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 81-07-2
Molecular Formula C7H5NO3S
Molecular Weight (g/mol) 183.18
InChI Key CVHZOJJKTDOEJC-UHFFFAOYSA-N
IUPAC Name 2,3-dihydro-1λ⁶,2-benzothiazole-1,1,3-trione
SMILES O=C1NS(=O)(=O)C2=CC=CC=C12

Specifications

CAS Max % 1
Ignition Residue 0.002
Loss on Drying 0.01
Melting Range 226-230°C
Packaging Poly Pail
Quantity 2.5 kg
Percent Purity 99 to 101%
Grade NF
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.