Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Saccharomyces cerevisiae MHT1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5117504
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5117504 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5117504 Supplier Invitrogen™ Supplier No. PA5117504
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody has been tested in direct-ELISA.

Homocysteine S-methyltransferase involved in the conversion of S-adenosylmethionine (AdoMet) to methionine to control the methionine/AdoMet ratio. Converts also S-methylmethionine (SMM) to methionine.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Saccharomyces cerevisiae MHT1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene MHT1
Gene Accession No. Q12525
Gene Alias Hcy S-methyltransferase 1; homocysteine methyltransferase 1; Homocysteine S-methyltransferase 1; L0552; MHT1; S-adenosylmethionine-homocysteine S-methyltransferase MHT1; S-methylmethionine; S-methylmethionine:homocysteine methyltransferase 1; SMM; SMM:Hcy S-methyltransferase 1; YLL062C
Gene Symbols MHT1
Host Species Rabbit
Immunogen Recombinant Saccharomyces cerevisiae Homocysteine S-methyltransferase 1 protein (1-324aa).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 850664
Target Species Yeast
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.