Learn More
Invitrogen™ Saccharomyces cerevisiae Polyclonal Antibody

Description
PA1-26103 detects Saccharomyces cerevisiae from yeast samples.
Specifications
Specifications
| Antigen | Saccharomyces cerevisiae |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.1% sodium azide; pH 7.2 |
| Gene Alias | S. cerevisiae |
| Host Species | Rabbit |
| Immunogen | Whole intact cells of S. cerevisiae. |
| Purification Method | Protein A |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.