Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Saccharomyces cerevisiae Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA17243
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA17243 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA17243 Supplier Invitrogen™ Supplier No. PA17243
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PA1-7243 detects the Saccharomyces cerevisiae. Cross reactivity with other species is not known. PA1-7243 has been successfully used in Western blot procedures. The PA1-7243 immunogen is whole cells of S. cerevisiae. Recommended short term storage (less than 6 months) is at 4°C. For longer term storage, aliquot and freeze.

Saccharomyces cerevisiae also known as baker's yeast, is a genus of ascomycetes. They are normally diploid unicellular fungi that reproduce asexually by budding. Asci, containing four haploid ascospores, develop directly from the diploid vegetative cells by meiosis. After germination of the ascospores the haploid cells can reproduce vegetatively, or haploid cells of different mating type can fuse to form a diploid zygote. Most laboratory strains used are, in contrast to wild type yeasts, stable haploids.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Saccharomyces cerevisiae
Applications Western Blot
Classification Polyclonal
Concentration 5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.2
Gene Alias S. cerevisiae
Host Species Rabbit
Immunogen Whole cells of S. cerevisiae.
Purification Method IgG fraction
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Yeast
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.