Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Saccharomyces cerevisiae PPX1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5117580
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5117580 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5117580 Supplier Thermo Scientific Supplier No. PA5117580
Add to Cart
Add to Cart

Saccharomyces cerevisiae PPX1 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen Saccharomyces cerevisiae PPX1
Applications Western Blot
Classification Polyclonal
Concentration 4 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene PPX1
Gene Accession No. P38698
Gene Alias ExopolyPase; Exopolyphosphatase; Metaphosphatase; PPX1; Ppx1p; YHR201C
Gene Symbols PPX1
Host Species Rabbit
Immunogen Recombinant Saccharomyces cerevisiae Exopolyphosphatase protein (1-397aa).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 856608
Target Species Yeast
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.